Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lactic Acid Solution

Catalog No. 252476500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-247-6500G 500 g
25-247-61KG -
25-247-625KG -
25-247-6100G -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 25-247-6500G Supplier Sigma Aldrich Supplier No. 252476500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lactic acid solution has been used in the preparation of poly(lactic acid) (PLA)/lactic acid (LA) blends to enhance the biodegradability rate of PLA.Contains varying amounts of intermolecular esterification products.

Specifications

CAS 50-21-5
Density 1.209 g/mL at 25°C
Quantity 500 g
Refractive Index n20/D 1.4251
Molecular Formula C3H6O3
Linear Formula CH3CH(OH)CO2H
MDL Number MFCD00004520
Molecular Weight (g/mol) 90.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.