Learn More
Medchemexpress LLC Lactic acid (Standard) | 50-21-5 | 91.1% | 25 MG
SDP
Catalog No. 5000413541
Encompass_Preferred
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product, linking glycolysis and oxidative phosphorylation. It is considered an oncometabolite, plays an immune protective role in anti-tumor immunity, and exhibits antimicrobial activity. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS techniques.
  • Activator of hydroxycarboxylic acid receptor 1 (HCAR1).
  • Epigenetic modulator that induces lactylation of lysine residues.
  • Glycolysis end-product.
  • Considered an oncometabolite.
  • Plays an immune protective role in anti-tumor immunity.
  • Exhibits antimicrobial activity, suitable as a food preservative.
Catalog No. 50-004-13541 Supplier Medchemexpress LLC Supplier No. 25MGHYB2227R
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product, linking glycolysis and oxidative phosphorylation. It is considered an oncometabolite, plays an immune protective role in anti-tumor immunity, and exhibits antimicrobial activity. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS techniques.
  • Activator of hydroxycarboxylic acid receptor 1 (HCAR1).
  • Epigenetic modulator that induces lactylation of lysine residues.
  • Glycolysis end-product.
  • Considered an oncometabolite.
  • Plays an immune protective role in anti-tumor immunity.
  • Exhibits antimicrobial activity, suitable as a food preservative.