Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Lactic Acid, USP (Racemic) (Dietary Supplement Grade)
SDP

Catalog No. 14185653 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-653 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-653 Supplier Medisca Supplier No. 056509
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lactic Acid, USP (Racemic) (Dietary Supplement Grade) is a colorless or yellowish, practically odorless, syrupy liquid that is miscible with both water and with alcohol.

Lactic Acid, USP (Racemic) is a colorless or yellowish, practically odorless, syrupy liquid that is miscible with both water and with alcohol. It is hygroscopic.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Specifications

Chemical Name or Material Lactic Acid, USP (Racemic) (Dietary Supplement Grade)
Packaging Bottle, 1000mL, Amber Glass, Boston Round, 33-430
CAS 50-21-5
Type Drug Standard
Molecular Weight (g/mol) 90.078
Quantity 1 L
Percent Purity 88 to 92.0 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.