Learn More
TRIBIOSCIENCE INC Lactic Lactate Fluorometric Assay 100 Tests
SDP
Catalog No. NC3942479
Encompass_Preferred
Tribioscience’s Lactate Fluorometric Assay Kit is a simple, sensitive, and fast assay that measures the total amount of L-Lactate in biological samples. Lactate is oxidized by lactate oxidase to pyruvate and hydrogen peroxide. Horseradish peroxidase (HRP) catalyzes the reaction between hydrogen peroxide and the specific probe to yield a final product measured by the fluorometric method at Ex/Em=530/590 nm.
Catalog No. NC3942479 Supplier TRIBIOSCIENCE INC Supplier No. TBS2061
Tribioscience’s Lactate Fluorometric Assay Kit is a simple, sensitive, and fast assay that measures the total amount of L-Lactate in biological samples. Lactate is oxidized by lactate oxidase to pyruvate and hydrogen peroxide. Horseradish peroxidase (HRP) catalyzes the reaction between hydrogen peroxide and the specific probe to yield a final product measured by the fluorometric method at Ex/Em=530/590 nm.