Learn More
ATCC Lacticaseibacillus casei; Strain 03
Catalog No. 502382938
Encompass
Lacticaseibacillus casei strain 03 was isolated from dairy products. This whole-genome sequenced bacterial type strain produces acetoin and diacetyl.
Catalog No. 50-238-2938 Supplier ATCC Supplier No. 393
Lacticaseibacillus casei strain 03 was isolated from dairy products. This whole-genome sequenced bacterial type strain produces acetoin and diacetyl.