Learn More
ATCC Lactiplantibacillus plantarum
Supplier:  ATCC 8014
Encompass
Lactiplantibacillus plantarum strain 17-5 is a whole-genome sequenced bacterium known to produce lactic acid. This strain is recommended as a test microorganism for USP <441> and USP <91>.
Catalog No. 50-238-3364
 Restricted Item