Learn More
Cayman Chemical Lactisole, 150436-68-3, 5 g
SDP
Supplier:  Cayman Chemical 186575
Encompass_Preferred
An antagonist of sweet taste receptors that blocks the activation of T1R3 by natural and synthetic sweeteners and increases the inhibition of T1R2 by umami compounds; used to explore the roles of these receptors in diverse pathways
Catalog No. NC1193967