Learn More
Medchemexpress LLC Lactitol | 585-86-4 | MFCD00079407 | 50 MG
SDP
Catalog No. 5000417646
Encompass_Preferred
Lactitol (D-Lactitol) is a non-absorbable disaccharide used in research. It functions by modifying gut microbiota, specifically reducing harmful populations while increasing beneficial ones, and is applied in studies related to portosystemic encephalopathy and chronic constipation.
  • Reduces populations of *Bacteroides*, *Clostridium*, coliforms, and *Eubacterium*
  • Increases populations of *Lactobacillus* and *Streptococcus*
  • Utilized in research for portosystemic encephalopathy
  • Utilized in research for chronic constipation
Catalog No. 50-004-17646 Supplier Medchemexpress LLC Supplier No. HYN710450MG
Lactitol (D-Lactitol) is a non-absorbable disaccharide used in research. It functions by modifying gut microbiota, specifically reducing harmful populations while increasing beneficial ones, and is applied in studies related to portosystemic encephalopathy and chronic constipation.
  • Reduces populations of *Bacteroides*, *Clostridium*, coliforms, and *Eubacterium*
  • Increases populations of *Lactobacillus* and *Streptococcus*
  • Utilized in research for portosystemic encephalopathy
  • Utilized in research for chronic constipation