Learn More
Selleck Chemical LLC Lactitol monohydrate S5282-25mg
SDP
Catalog No. 503112245
Encompass_Preferred
Lactitol is an artificial sugar alcohol currently used as a bulk sweetener in calorie-controlled foods
Catalog No. 50-311-2245 Supplier Selleck Chemical LLC Supplier No. S528225MG
Lactitol is an artificial sugar alcohol currently used as a bulk sweetener in calorie-controlled foods