Learn More
Medchemexpress LLC Lacto-N-biose I | 50787-09-2 | MFCD00036714 | 98.0% | C14H25NO11 | 1 MG
SDP
Catalog No. 5000355631
Encompass_Preferred
Lacto-N-biose I (Galβ1-3GlcNAc), an endogenous metabolite, functions as an acceptor for the α1,2-fucosyltransferase enzyme found in Helicobacter pylori. It is intended for research use only and is not sold to patients.
  • Endogenous metabolite
  • Acceptor for α1,2-fucosyltransferase enzyme
  • Acts on Helicobacter pylori
  • For research use only
Catalog No. 50-003-55631 Supplier Medchemexpress LLC Supplier No. HY1414881MG
Lacto-N-biose I (Galβ1-3GlcNAc), an endogenous metabolite, functions as an acceptor for the α1,2-fucosyltransferase enzyme found in Helicobacter pylori. It is intended for research use only and is not sold to patients.
  • Endogenous metabolite
  • Acceptor for α1,2-fucosyltransferase enzyme
  • Acts on Helicobacter pylori
  • For research use only