Learn More
Medchemexpress LLC Lacto-N-fucopentaose I | 7578-25-8 | MFCD00792849 | 98.0% | 853.77 g/mol | C32H55NO25 | 1 MG
SDP
Catalog No. 5000300099
Encompass_Preferred
Lacto-N-fucopentaose I is a human milk oligosaccharide used as a research reagent in studies of antiviral and antibacterial activity, viral adsorption inhibition, cell cycle regulation, reactive oxygen species, apoptosis, and modulation of intestinal microbiota and immune development. It is provided as a purified, water-soluble oligosaccharide suitable for biochemical and cell-based assays.
  • High purity suitable for analytical and biological studies.
  • Water-soluble for easy preparation of stock solutions.
  • Useful in studies of viral adsorption and capsid protein interactions.
  • Supports investigation of cell cycle regulation and apoptosis.
  • Stable under recommended storage for reproducible results.
Catalog No. 50-003-00099 Supplier Medchemexpress LLC Supplier No. HYW0198061MG
Lacto-N-fucopentaose I is a human milk oligosaccharide used as a research reagent in studies of antiviral and antibacterial activity, viral adsorption inhibition, cell cycle regulation, reactive oxygen species, apoptosis, and modulation of intestinal microbiota and immune development. It is provided as a purified, water-soluble oligosaccharide suitable for biochemical and cell-based assays.
  • High purity suitable for analytical and biological studies.
  • Water-soluble for easy preparation of stock solutions.
  • Useful in studies of viral adsorption and capsid protein interactions.
  • Supports investigation of cell cycle regulation and apoptosis.
  • Stable under recommended storage for reproducible results.