Learn More
Medchemexpress LLC Lacto-N-fucopentaose II (LNFP II) | 21973-23-9 | 1 MG
SDP
Catalog No. 5000419940
Encompass_Preferred
Lacto-N-fucopentaose II (LNFP II) is a sialyl-Lewis, a hapten of human Lewis bloodgroup determinant. It is also a monosialo-ganglioside/glycolipid, and its sialyl derivative, CA 19-9, is recognized as a molecular tumor marker for biliopancreatic malignancy. This product has a purity of 98.0% and appears as a white to off-white solid, intended for research use only.
  • Sialyl-Lewis, a hapten of human Lewis bloodgroup determinant
  • Monosialo-ganglioside/glycolipid
  • Sialyl derivative, CA 19-9, is a molecular tumor marker for biliopancreatic malignancy
  • Purity of 98.0%
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-19940 Supplier Medchemexpress LLC Supplier No. HYN75751MG
Lacto-N-fucopentaose II (LNFP II) is a sialyl-Lewis, a hapten of human Lewis bloodgroup determinant. It is also a monosialo-ganglioside/glycolipid, and its sialyl derivative, CA 19-9, is recognized as a molecular tumor marker for biliopancreatic malignancy. This product has a purity of 98.0% and appears as a white to off-white solid, intended for research use only.
  • Sialyl-Lewis, a hapten of human Lewis bloodgroup determinant
  • Monosialo-ganglioside/glycolipid
  • Sialyl derivative, CA 19-9, is a molecular tumor marker for biliopancreatic malignancy
  • Purity of 98.0%
  • White to off-white solid appearance
  • For research use only