Learn More
ALPHA BIOSCIENCES Lactobacillus MRS Broth w/o Dextrose, 500g
Supplier:  ALPHA BIOSCIENCES L12104M500G
Encompass_Preferred
Lactobacilli MRS Broth, also referred to as Lactobacillus MRS Broth or LMRS Broth, is an agar medium used for the selective isolation and enumeration of Lactobacilli from various sources which include feces and dairy. Beef Extract and Yeast Extract provide amino acids, peptides, carbohydrates, vitamins, and minerals. Sodium Acetate and Ammonium Citrate serve as both selective agents and energy sources. Sodium phosphate acts as pH buffer, while Magnesium and Manganese Sulfates provide divalent cations essential for bacterial metabolism. Tween 80 is a surfactant to facilitate nutrient uptake by the Lactobacilli. Dextrose is not included in this version of the formulation to provide the user with the freedom to incorporate a desired carbohydrate for additional selectivity.
Catalog No. NC1646815