Learn More
ALPHA BIOSCIENCES Lactobacilli MRS Agar | 500g
Catalog No. NC1729537
Encompass_Preferred
Lactobacilli MRS Agar, also referred to as Lactobacillus MRS Agar or LMRS Agar, is an agar medium used for the selective isolation and enumeration of Lactobacilli from various sources which include feces and dairy. Proteose Peptone, Beef Extract, and Yeast Extract provide nitrogenous sources, carbohydrates, vitamins, and minerals. Dextrose is the main fermentable carbohydrate. Sodium Acetate and Ammonium Citrate serve as both selective agents and energy sources. Potassium Phosphate acts as pH buffer, while Magnesium and Manganese Sulfates provide cations essential for bacterial metabolism. Tween 80 is a surfactant to facilitate nutrient uptake by the Lactobacilli. Agar is the gelifying agent.
Catalog No. NC1729537 Supplier ALPHA BIOSCIENCES Supplier No. L12103500G
Lactobacilli MRS Agar, also referred to as Lactobacillus MRS Agar or LMRS Agar, is an agar medium used for the selective isolation and enumeration of Lactobacilli from various sources which include feces and dairy. Proteose Peptone, Beef Extract, and Yeast Extract provide nitrogenous sources, carbohydrates, vitamins, and minerals. Dextrose is the main fermentable carbohydrate. Sodium Acetate and Ammonium Citrate serve as both selective agents and energy sources. Potassium Phosphate acts as pH buffer, while Magnesium and Manganese Sulfates provide cations essential for bacterial metabolism. Tween 80 is a surfactant to facilitate nutrient uptake by the Lactobacilli. Agar is the gelifying agent.