Learn More
NEOGEN Lactobacilli MRS Agar, 5 kg
SDP
Catalog No. 502015256
Encompass_Preferred
Lactobacilli MRS Agar is used for the cultivation of lactobacilli and is not intended for use in the diagnosis of disease or other conditions in humans.MRS Agar is a medium for the cultivation and enumeration of Lactobacillus spp. Originally developed in 1960 by de Man, Rogosa & Sharpe, the medium is suitable for most lactic acid bacteria and is intended as a substitute for Tomato Juice Agar. When acidified to pH 5.4 MRS Agar can be used to enumerate Lactobacillus bulgaricus in yogurts.Nutrition is provided by a mixture of carefully selected peptones, glucose, beef & yeast extracts while Tween 80, magnesium and manganese sulfates act as growth stimulants. Selectivity against streptococci & molds is provided by ammonium citrate and sodium acetate. Used at low pH, ammonium citrate allows growth of lactobacilli while inhibiting a number of other microorganism groups.For more information visit https://www.neogen.com/solutions/Microbiology/lactobacilli-mrs-agar
Catalog No. 50-201-5256 Supplier NEOGEN Supplier No. 700003066
Lactobacilli MRS Agar is used for the cultivation of lactobacilli and is not intended for use in the diagnosis of disease or other conditions in humans.MRS Agar is a medium for the cultivation and enumeration of Lactobacillus spp. Originally developed in 1960 by de Man, Rogosa & Sharpe, the medium is suitable for most lactic acid bacteria and is intended as a substitute for Tomato Juice Agar. When acidified to pH 5.4 MRS Agar can be used to enumerate Lactobacillus bulgaricus in yogurts.Nutrition is provided by a mixture of carefully selected peptones, glucose, beef & yeast extracts while Tween 80, magnesium and manganese sulfates act as growth stimulants. Selectivity against streptococci & molds is provided by ammonium citrate and sodium acetate. Used at low pH, ammonium citrate allows growth of lactobacilli while inhibiting a number of other microorganism groups.For more information visit https://www.neogen.com/solutions/Microbiology/lactobacilli-mrs-agar