Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Lactobacilli MRS Agar
SDP

Catalog No. 23001778 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Lactobacillus delbrueckii, Lactobacillus fermentum, Lactobacillus acidophilus, Lactobacillus species
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-778 Lactobacillus delbrueckii, Lactobacillus fermentum, Lactobacillus acidophilus, Lactobacillus species
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-778 Supplier Hardy Diagnostics™ Supplier No. G117
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the isolation and enumeration of lactobacillus app.

  • Lactobacilli MRS Agar with Cycloheximide is useful for inhibiting fungi that may be in a sample
  • 10 per package

Specifications

Appearance Slightly Opalescent
Color Medium Amber
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Tween™ 80, Sodium Acetate, Ammonium Citrate, Magnesium Sulfate, Manganese Sulfate
Detects Lactobacillus spp.
Form Agar
Format Plate
Manufacturing Origin United States
Media Type Lactobacilli MRS Agar
Product Type Agar Plate
Target Organism Class Multidrug-resistant, Gram-negative Organisms
Test Type Microbial Detection
Fill Volume 18 mL
Formulation Dextrose - 20.0gm, Peptic Digest of Animal Tissue - 10.0gm, Beef Extract - 10.0gm, Yeast Extract - 5.0gm, Sodium Acetate - 5.0gm, Dipotassium Phosphate - 2.0gm, Ammonium Citrate - 2.0gm, Tween 80 - 1.0gm, Magnesium Sulfate - 0.1gm, Manganese Sulfate - 0.05gm, Agar - 15.0gm
For Use With (Application) Isolation, enumeration, and cultivation of Lactobacillus spp.
Incubation Conditions 35°C
Incubation Time 24 to 72 hr.
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Lactobacillus delbrueckii, Lactobacillus fermentum, Lactobacillus acidophilus, Lactobacillus species
Protein Source Animal Tissue, Beef, Yeast
Source Animal Tissue, Beef, Yeast
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.