Learn More
NEOGEN Lactobacilli MRS Broth, 10 kg
SDP
Catalog No. 502015386
Encompass_Preferred
Lactobacilli MRS Broth is used for the cultivation of lactobacilli and is not intended for use in the diagnosis of disease or other conditions in humans.Lactobacilli MRS broth was originally developed in 1960 by de Man, Rogosa & Sharpe, the medium can be used for confirmatory tests on organisms isolated on MRS Agar. The medium can also be used for enumeration by the Miles and Misra technique.Nutrition is provided by a mixture of carefully selected peptones, glucose, beef and yeast extracts while Tween 80, magnesium and manganese sulfates act as growth stimulants. Selectivity against streptococci & molds is provided by ammonium citrate and sodium acetate.For more information visit https://www.neogen.com/solutions/Microbiology/lactobacilli-mrs-broth
Catalog No. 50-201-5386 Supplier NEOGEN Supplier No. 700003191
Lactobacilli MRS Broth is used for the cultivation of lactobacilli and is not intended for use in the diagnosis of disease or other conditions in humans.Lactobacilli MRS broth was originally developed in 1960 by de Man, Rogosa & Sharpe, the medium can be used for confirmatory tests on organisms isolated on MRS Agar. The medium can also be used for enumeration by the Miles and Misra technique.Nutrition is provided by a mixture of carefully selected peptones, glucose, beef and yeast extracts while Tween 80, magnesium and manganese sulfates act as growth stimulants. Selectivity against streptococci & molds is provided by ammonium citrate and sodium acetate.For more information visit https://www.neogen.com/solutions/Microbiology/lactobacilli-mrs-broth