Learn More
Bioworld Lactobacilli MRS Broth, 500 g
Catalog No. 50254308
Encompass_Preferred
Lactobacilli MRS BrothFor the cultivation of Lactobacillus species from oral, fecal and other specimen sourcespH: 6.5Working Concentration: 55 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: amber brothSoluble in boiling waterComposition:Peptic Digest of Animal Tissue Beef Extract Yeast Extract Dextrose Sodium Acetate Tween 80 Disodium Phosphate Ammonium Citrate Magnesium Sulfate Manganese Sulfate
Catalog No. 50-254-308 Supplier Bioworld Supplier No. 306200311
Lactobacilli MRS BrothFor the cultivation of Lactobacillus species from oral, fecal and other specimen sourcespH: 6.5Working Concentration: 55 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: amber brothSoluble in boiling waterComposition:Peptic Digest of Animal Tissue Beef Extract Yeast Extract Dextrose Sodium Acetate Tween 80 Disodium Phosphate Ammonium Citrate Magnesium Sulfate Manganese Sulfate