Learn More
ATCC Komagataeibacter rhaeticus ;Strain: iGEM
Catalog No. 502381094
Encompass
The depositor claims that this strain is a cellulose producer and a nitrogen fixer, and is resistant to ampicillin, kanamycin, and chloramphenicol.
Catalog No. 50-238-1094 Supplier ATCC Supplier No. BAA2831
The depositor claims that this strain is a cellulose producer and a nitrogen fixer, and is resistant to ampicillin, kanamycin, and chloramphenicol.