Learn More
ATCC Lactobacillus crispatus
Catalog No. 502384406
Encompass
Lactobacillus crispatus strain VPI 7635 was isolated from urine. This whole-genome sequenced bacterium is in homology group A2 and is propagated in a 5% CO2 atmosphere.
Catalog No. 50-238-4406 Supplier ATCC Supplier No. 33197
Add to Cart
Add to Cart
Lactobacillus crispatus strain VPI 7635 was isolated from urine. This whole-genome sequenced bacterium is in homology group A2 and is propagated in a 5% CO2 atmosphere.