Learn More
ATCC Lactobacillus delbrueckii; subsp. lactis; Strain L110
Supplier:  ATCC 12315
Encompass
Lactobacillus delbrueckii subsp. lactis strain L110 was isolated from dairy products. This bacterial type strain is propagated aerobically in lactobacilli MRS medium.
Catalog No. 50-238-3737