Learn More
ATCC Lactobacillus delbrueckii subsp. delbrueckii strain 730 (ATCC 9649™)
Catalog No. 502382653
Encompass
Lactobacillus delbrueckii subsp. delbrueckii strain 730 (ATCC 9649™) is a whole-genome sequenced type strain isolated from sour grain mash. It produces D(-)lactic acid and is used in food production research and bioinformatics.
Recommended companion product:
Quantitative Genomic DNA (Fisher Cat. No. 50-238-3915, ATCC 9649DQ™), ideal for qPCR assay development and microbial load quantification.
Catalog No. 50-238-2653 Supplier ATCC Supplier No. 9649
Lactobacillus delbrueckii subsp. delbrueckii strain 730 (ATCC 9649™) is a whole-genome sequenced type strain isolated from sour grain mash. It produces D(-)lactic acid and is used in food production research and bioinformatics.
Recommended companion product:
Quantitative Genomic DNA (Fisher Cat. No. 50-238-3915, ATCC 9649DQ™), ideal for qPCR assay development and microbial load quantification.