Learn More
ATCC Lactobacillus fermentum; Strain 36
Supplier:  ATCC 9338
Encompass
Lactobacillus fermentum strain 36 can be used in assays of alanine, histidine, pyrithiamine, and thiamine.
Catalog No. 50-238-2693
 Restricted Item