Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Bispebjerg
Catalog No. 502381230
Encompass
Bacteriophage host. Used as a control strain for the detection of Salmonella and in the examination of dairy products.
Catalog No. 50-238-1230 Supplier ATCC Supplier No. 9842
Add to Cart
Add to Cart
Bacteriophage host. Used as a control strain for the detection of Salmonella and in the examination of dairy products.