Learn More
ATCC Mycobacterium tuberculosis; genomic DNA
Catalog No. 502381198
Encompass
Genomic DNA isolated from Mycobacterium tuberculosis Strain TMC 102 [H37Rv]. This bacterial strain is also available as ATCC Catalog No. 27294™.
Catalog No. 50-238-1198 Supplier ATCC Supplier No. 27294D2
Add to Cart
Add to Cart
Genomic DNA isolated from Mycobacterium tuberculosis Strain TMC 102 [H37Rv]. This bacterial strain is also available as ATCC Catalog No. 27294™.