Learn More
ATCC Levilactobacillus brevis
Catalog No. 502379821
Encompass
Bacterial strain isolated from green fermenting Sevillano variety olives. Growth of this product is best in broth culture or on biphasic slants.
Catalog No. 50-237-9821 Supplier ATCC Supplier No. 8287
Bacterial strain isolated from green fermenting Sevillano variety olives. Growth of this product is best in broth culture or on biphasic slants.