Learn More
ATCC Lactobacillus johnsonii
Catalog No. 502382062
Encompass
Lactobacillus johnsonii strain VPI 7960 is a whole-genome sequenced bacterial type strain that was isolated from human blood in Belgium.
Catalog No. 50-238-2062 Supplier ATCC Supplier No. 33200
Lactobacillus johnsonii strain VPI 7960 is a whole-genome sequenced bacterial type strain that was isolated from human blood in Belgium.