Learn More
ATCC Lactobacillus delbrueckii subsp. lactis
Catalog No. 502383638
Encompass
Lactobacillus delbrueckii subsp. lactis strain 313 is a whole-genome sequenced strain. This bacterial culture is recommended as a test microorganism for USP <171> Vitamin B12 Activity Assay.
Catalog No. 50-238-3638 Supplier ATCC Supplier No. 7830
Lactobacillus delbrueckii subsp. lactis strain 313 is a whole-genome sequenced strain. This bacterial culture is recommended as a test microorganism for USP <171> Vitamin B12 Activity Assay.