Learn More
ATCC Lactobacillus delbrueckii subsp. lactis; 6 x 0.3mL
Catalog No. 502382691
Encompass
Lactobacillus delbrueckii subsp. lactis strain 313 (ATCC 7830-MINI-PACK) consists of 6 ready-to-use vials of ATCC 7830 frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-2691 Supplier ATCC Supplier No. 7830MINIPACK
Lactobacillus delbrueckii subsp. lactis strain 313 (ATCC 7830-MINI-PACK) consists of 6 ready-to-use vials of ATCC 7830 frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.