Learn More
Alkali Scientific Lactobacillus Mrs Agar w/Cycloheximide & Maltose 10mL Plate, 10/Pk
SDP
Catalog No. 5000662172
Encompass_Preferred
Lactobacillus MRS Agar with Cycloheximide and Maltose is a selective medium developed for the isolation and enumeration of lactic acid bacteria in fermentation and brewing environments. The formulation is based on the classic MRS nutrient system that supports the growth of Lactobacillus and Pediococcus species while maintaining optimal pH and nutrient balance. Cycloheximide is incorporated as an antifungal agent to suppress yeast growth commonly present in beer, wort, and wine samples. Maltose serves as a preferred carbohydrate source for many lactic acid bacteria, improving recovery of brewing contaminants. This medium allows reliable detection of bacterial spoilage organisms in fermentation processes and is widely used in brewery quality control laboratories.
Catalog No. 50-006-62172 Supplier Alkali Scientific Supplier No. LMLP00
Lactobacillus MRS Agar with Cycloheximide and Maltose is a selective medium developed for the isolation and enumeration of lactic acid bacteria in fermentation and brewing environments. The formulation is based on the classic MRS nutrient system that supports the growth of Lactobacillus and Pediococcus species while maintaining optimal pH and nutrient balance. Cycloheximide is incorporated as an antifungal agent to suppress yeast growth commonly present in beer, wort, and wine samples. Maltose serves as a preferred carbohydrate source for many lactic acid bacteria, improving recovery of brewing contaminants. This medium allows reliable detection of bacterial spoilage organisms in fermentation processes and is widely used in brewery quality control laboratories.