Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Lactobacillus MRS Broth Powder, 500 Grams
SDP

Catalog No. 50488730
Encompass_Preferred

Recommended for the selective isolation and enumeration of lactobacilli.

Ingredients:

Peptic Digest of Animal Tissue: 10.0g
Beef Extract: 10.0g
Yeast Extract: 5.0g
Ammonium Citrate: 2.0g
Dextrose: 20.0g
Polysorbate 80:1.0g
Sodium acetate: 5.0g
Magnesium Sulfate: 0.1g
Manganese Sulfate: 0.05g
Sodium Phosphate, Dibasic: 2.0g

Catalog No. 50-488-730 Supplier Research Products International Corp Supplier No. L11000500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended for the selective isolation and enumeration of lactobacilli.

Ingredients:

Peptic Digest of Animal Tissue: 10.0g
Beef Extract: 10.0g
Yeast Extract: 5.0g
Ammonium Citrate: 2.0g
Dextrose: 20.0g
Polysorbate 80:1.0g
Sodium acetate: 5.0g
Magnesium Sulfate: 0.1g
Manganese Sulfate: 0.05g
Sodium Phosphate, Dibasic: 2.0g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.