Learn More
Zeptometrix Lactobacillus plantarum Z524, Titered
SDP
Catalog No. 22158414 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-414 1 mL
1 options
Catalog No. 22-158-414 Supplier Zeptometrix Supplier No. 0804436
Add to Cart
Add to Cart
Infectious, titered microorganisms can be utilized for analytical testing during assay development, including limit of detection (LOD) and cross-reactivity studies. When used as a control for nucleic acid tests, the same protocols as those used to extract and amplify clinical specimens should be employed.
Each frozen aliquot contains 1 mL of a pure, titered culture of Lactobacillus plantarum. The identification of this organism was confirmed using whole genome sequencing. The purity of the culture was monitored by Gram staining and by additional culturing. The titer was performed on a minimum of two aliquots after freezing. The freezing medium contains 15% glycerol as a cryoprotectant. Please see the Certificate of Analysis for the specific freezing medium used.
Specifications
Biosafety Level 1
Certifications/Compliance ISO 13485
Product Type Microorganism
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
Regulatory Status RUO
The purchase of infectious microorganisms from ZeptoMetrix requires a Material Transfer Agreement (MTA).