Learn More
ATCC Lactobacillus sakei; subsp. sakei
Catalog No. 502383490
Encompass
Lactobacillus sakei subsp. sakei strain T.S [K. Kitahara 37] was isolated from a starter batch of sake. This bacterial type strain can be used in media testing.
Catalog No. 50-238-3490 Supplier ATCC Supplier No. 15521
Add to Cart
Add to Cart
Lactobacillus sakei subsp. sakei strain T.S [K. Kitahara 37] was isolated from a starter batch of sake. This bacterial type strain can be used in media testing.