Learn More
ATCC Thermus thermophilus; Strain HB8
Catalog No. 502379605
Encompass
Type strain. Used in enzyme studies for phosphoglucomutase, phosphofructokinase, and fructose 1,6-diphosphatase. Resistant to heavy metal ions. Produces thermostable D-xylose isomerase.and restriction endonuclease TthHB8I. Genome sequenced strain.
Catalog No. 50-237-9605 Supplier ATCC Supplier No. 27634
Type strain. Used in enzyme studies for phosphoglucomutase, phosphofructokinase, and fructose 1,6-diphosphatase. Resistant to heavy metal ions. Produces thermostable D-xylose isomerase.and restriction endonuclease TthHB8I. Genome sequenced strain.