Learn More
Medchemexpress LLC HY-N7059 100mg Medchemexpress, Lactobionic acid CAS:96-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7059100MG
Encompass_Preferred
Medchemexpress, HY-N7059 100mg Lactobionic acid CAS:96-82-2 Lactobionic acid is a bionic acid naturally found in the Caspian Sea yogurt and chemically constituted of a gluconic acid bonded to a galactose. Lactobionic acid has antioxidant, antimicrobial, chelating, stabilizer, acidulant, and moisturizing properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0494