Learn More
AG Scientific Inc Lactobionic Acid, 500 G
Catalog No. 502236996
Encompass_Preferred
An inhibitor of matrix metalloproteinases (MMP).CAS Number: 96-82-2Molecular Weight: 358.3 DaChemical Formula: C12H22O12Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6996 Supplier AG Scientific Inc Supplier No. L2579500GM
An inhibitor of matrix metalloproteinases (MMP).CAS Number: 96-82-2Molecular Weight: 358.3 DaChemical Formula: C12H22O12Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.