Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactobionic Acid, NF, EP, 500 g, Spectrum Chemical
SDP

Catalog No. 18615534 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-615-534 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-615-534 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1604500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lactobionic Acid, NF, EP is also known as Galactosylgluconic acid. It is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The EP grade indicates it is grade suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Weight (g/mol) 358.3 g/mol

Specifications

Quantity 500 g
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Lactobionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.