Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactobionic Acid, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18620954 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
100 g
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-954 5 g
18-620-955 100 g
18-620-956 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-954 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L10175GM
Add to Cart
Add to Cart

Synonym: Galactosylgluconic Acid, 4-O-Beta-D-Galactopyranosyl-D-Gluconic Acid, 4-(Beta-D-Galactosido)-D-Gluconic Acid

Lactobionic Acid, also known as Galactosylgluconic acid, is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical™ manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 96-82-2
Molecular Weight (g/mol) 358.3 g/mol

Specifications

Quantity 5 g
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Lactobionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.