Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactobionic Acid, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18620956 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
100 g
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-956 10 kg
18-620-954 5 g
18-620-955 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-956 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L101710KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: Galactosylgluconic Acid, 4-O-Beta-D-Galactopyranosyl-D-Gluconic Acid, 4-(Beta-D-Galactosido)-D-Gluconic Acid

Lactobionic Acid, also known as Galactosylgluconic acid, is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical™ manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 96-82-2
Molecular Weight (g/mol) 358.3 g/mol

Specifications

Packaging Drum
Quantity 10 kg
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Lactobionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.