Learn More
Medchemexpress LLC HY-P1791B 5mg , Lactoferrin (17-41) (acetate) CAS: Purity:>98%
SDP
Catalog No. 502261988
Encompass_Preferred
HY-P1791B 5mg Lactoferrin (17-41) (acetate) CAS: Lactoferrin 17-41 (Lactoferricin B) acetate, a peptide corresponding to residues 17-41 of bovine lactoferrin, has antimicrobial activity against a wide range of microorganisms, including Gram-positive and Gramnegative bacteria, viruses, protozoa, and fungi. Lactoferrin 17-41 acetate has antitumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1988 Supplier Medchemexpress LLC Supplier No. HYP1791B5MG
Add to Cart
Add to Cart
HY-P1791B 5mg Lactoferrin (17-41) (acetate) CAS: Lactoferrin 17-41 (Lactoferricin B) acetate, a peptide corresponding to residues 17-41 of bovine lactoferrin, has antimicrobial activity against a wide range of microorganisms, including Gram-positive and Gramnegative bacteria, viruses, protozoa, and fungi. Lactoferrin 17-41 acetate has antitumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.