Learn More
Sigma Aldrich Fine Chemicals Biosciences Lactoferrin from bovine milk, 146897-68-9, MFCD00131481, 50 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L950750MG
Encompass
≥85% (SDS-PAGE). Lactoferrin was used in the fractionation of lactoperoxidase and lactoferrin from bovine whey using a cation exchange membrane. It was used in the determination of lactoferrin and immunoglobulin G in animal milks by new immunosensors.
Catalog No. NC1043183