Learn More
Athens Research And Technology Inc LACTOFERRIN HU MILK 5MG
Supplier:  Athens Research And Technology Inc 16131201035MG
Encompass_Preferred
Lactoferrin, Human Milk: 5 MG; Frozen in 50 mM Tris-HCl, pH 8.0, with 0.2 mM NaCl; MW 82,400; EC= 1.46; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human milk in the Athens Research & Tech lab.
Catalog No. 50-489-845