Learn More
Bioss Lactoferrin Recombinant Monocl
SDP
Catalog No. 5000066465
Encompass_Preferred
Lactoferrin Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IF(ICC) IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66465 Supplier Bioss Supplier No. BSM62893R
Lactoferrin Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IF(ICC) IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol