Learn More
Med Vet International Lactophenol Cotton Blue, 4 oz, J0326B, Each
Supplier:  Med Vet International JOR326B
Encompass_Preferred
This stain is useful for fungal exams. It is used in combination with the 10 percent KOH (JOR326). First the KOH clears various debris; then the lactophenol cotton blue will stain the fungal elements a deep blue. 120ml.
Catalog No. 50-005-83008