Learn More
NEOGEN Lactose Broth, 5 kg
SDP
Catalog No. 502015169
Encompass_Preferred
Lactose Broth is used for the cultivation of Salmonella and coliform bacteria from food, dairy, and water products in a laboratory setting. Lactose Broth is not intended for use in the diagnosis of disease or other conditions in humans. Lactose Broth is frequently used as a pre-enrichment medium when testing foods and dairy products for Salmonella spp. In dried or processed foods, Salmonella species may be sub-lethally injured and in low numbers. The presence of other bacteria as well as components of the food sample may hinder growth and recovery of Salmonella. Pre-enrichment in a non-selective medium such as Lactose Broth allows for repair of cell damage, dilutes toxic or inhibitory substances, and provides a nutritional advantage to Salmonella over other bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/lactose-broth
Catalog No. 50-201-5169 Supplier NEOGEN Supplier No. 700002967
Lactose Broth is used for the cultivation of Salmonella and coliform bacteria from food, dairy, and water products in a laboratory setting. Lactose Broth is not intended for use in the diagnosis of disease or other conditions in humans. Lactose Broth is frequently used as a pre-enrichment medium when testing foods and dairy products for Salmonella spp. In dried or processed foods, Salmonella species may be sub-lethally injured and in low numbers. The presence of other bacteria as well as components of the food sample may hinder growth and recovery of Salmonella. Pre-enrichment in a non-selective medium such as Lactose Broth allows for repair of cell damage, dilutes toxic or inhibitory substances, and provides a nutritional advantage to Salmonella over other bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/lactose-broth