Learn More
R Biopharm Inc R-Biopharm Enzytec™ Lactose/D-Galactose Test Kit, UV Assay, AOAC Approved, 1 Test Kit
Catalog No. NC2212970
Encompass
The R-Biopharm Enzytec™ Generic Lactose/D-Galactose is a robust enzymatic test kit specifically designed for the accurate determination of lactose and D-galactose in foodstuffs and other materials. With an AOAC-approved method and high sensitivity using a UV assay at 340 nm, it ensures reliable quantitative results. Ideal for rigorous laboratory applications, its key advantages include minimal interference and validated standardization from international organizations.
This item is not returnable. View return policy
Catalog No. NC2212970 Supplier R Biopharm Inc Supplier No. E1213
The R-Biopharm Enzytec™ Generic Lactose/D-Galactose is a robust enzymatic test kit specifically designed for the accurate determination of lactose and D-galactose in foodstuffs and other materials. With an AOAC-approved method and high sensitivity using a UV assay at 340 nm, it ensures reliable quantitative results. Ideal for rigorous laboratory applications, its key advantages include minimal interference and validated standardization from international organizations.