Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Lactose Intolerance: Inheritance And Variation Of Traits
SCI_ED

Catalog No. S01997 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01997 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01997 Supplier Innovating Science™ Supplier No. IS3017
Add to Cart
Add to Cart

Lactose intolerance is common, but is actually a recessive genetic trait! Learn about variation, inheritance, and the role of DNA and proteins in expression of traits in this three-part laboratory activity!

Lactose intolerance is common, but is actually a recessive genetic trait! Humans and other mammals are only meant to drink milk as infants and as they grow, they lose the ability to digest lactose. As populations began to develop, the domestication of cows and drinking milk into adulthood became more common, leading to a tolerance for lactose. Learn about variation, inheritance, and the role of DNA and proteins in expression of traits in this three-part laboratory activity!

Specifications

Grade 6+
Class Size 15 Lab Groups
Product Type Enzyme Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.