Learn More
YSI 3P LifeScience LACTOSE MEMBRANES 4/PK
Catalog No. 50200119
Encompass
Lactose Membranes (galactose oxidase). Used to measure lactose or galactose on YSI 2700 and 2900 Series Analyzers and galactose on YSI 7100 Analyzers. NOTE: Requires overnight shipping.
Catalog No. 50-200-119 Supplier YSI 3P LifeScience Supplier No. 2702
Lactose Membranes (galactose oxidase). Used to measure lactose or galactose on YSI 2700 and 2900 Series Analyzers and galactose on YSI 7100 Analyzers. NOTE: Requires overnight shipping.