Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lactose, Monohydrate
SCI_ED

Catalog No. s25375 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25375 100 g Lab Grade
S25375A 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25375
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Color White
Quantity 100 g
Assay Percent Range 100% w/v
Linear Formula C12H22O11·H2O
Formula Weight 360.32
Grade Laboratory
Packaging Lab Grade
Density 1.525g/mL
Chemical Name or Material Lactose, Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.