Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactose, Monohydrate, BiotechGrade, Spectrum™ Chemical
SDP

Catalog No. 18603690 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-690 500 g Poly Bottle
18-603-691 2.5 kg Poly Bottle
18-603-692 12 kg Poly Bottle
18-603-693 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-690 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1037500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

L1037, 64044-51-5, C12H22O11·H2O

Spectrum™ Chemical Lactose, Monohydrate, BiotechGrade , also known as milk sugar, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
InChI Key WSVLPVUVIUVCRA-RJMJUYIDSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
SMILES O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 500 g
Grade Biotech
Loss on Drying 0.5%
Packaging Poly Bottle
Appearance Fine, white powder
Chemical Name or Material Lactose, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.